1-(2-Methylquinolin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

92793-500MG 92793-1G 92793-5G 92793-25G
77335-152EA 244.11 USD
77335-152 77359-246 77371-192 77391-666
1-(2-Methylquinolin-3-yl)ethanone ≥97%
1-(2-Methylquinolin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MW: 185.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09754368
CAS Number: 14208-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR