2-(4-Fluoro-2-methylphenyl)-2-oxoacetic acid ≥97%

Supplier: AOBChem USA

Warning

92550-500MG 92550-1G 92550-5G
77338-970EA 536.63 USD
77338-970 77359-244 77383-216
2-(4-Fluoro-2-methylphenyl)-2-oxoacetic acid ≥97%
2-(4-Fluoro-2-methylphenyl)-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃
MW: 182.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773209
CAS Number: 1896263-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR