2,4,6-trifluoro-3-isobutoxyphenol ≥97%

Supplier: AOBChem USA

Warning

92457-500MG 92457-1G 92457-5G 92457-25G
77335-150EA 120.51 USD
77335-150 77343-854 77371-190 77383-212
2,4,6-trifluoro-3-isobutoxyphenol ≥97%
2,4,6-trifluoro-3-isobutoxyphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34773203
CAS Number: 2901103-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR