1-(5-Bromo-2-fluorophenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

92449-500MG 92449-1G 92449-5G 92449-25G
77335-146EA 120.51 USD
77335-146 77343-852 77371-186 77383-210
1-(5-Bromo-2-fluorophenyl)cyclohexanol ≥97%
1-(5-Bromo-2-fluorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrFO
MW: 273.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23759082
CAS Number: 1515696-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR