(2-Fluoro-3-iodo-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

92430-1G 92430-5G 92430-25G
77341-116EA 71.07 USD
77341-116 77371-184 77383-208
(2-Fluoro-3-iodo-4-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-iodo-4-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₅F₄IS
MW: 336.088 g/mol
Storage Temperature: Ambient
CAS Number: 2705760-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR