(3-Bromophenyl)(5-Bromopyridin-3-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

92328-500MG 92328-1G 92328-5G 92328-25G
77335-144EA 182.31 USD
77335-144 77347-064 77371-182 77391-662
(3-Bromophenyl)(5-Bromopyridin-3-yl)methanone ≥97%
(3-Bromophenyl)(5-Bromopyridin-3-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₇Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD34773129
CAS Number: 2003106-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR