5-Chloro-4-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

92285-500MG 92285-1G 92285-5G
77336-126EA 375.95 USD
77336-126 77359-238 77383-206
5-Chloro-4-methylnicotinaldehyde ≥97%
5-Chloro-4-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD18258053
CAS Number: 1211515-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR