1-(3-Bromo-5-methoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

92186-500MG 92186-1G 92186-5G
77335-142EA 326.51 USD
77335-142 77359-234 77383-202
1-(3-Bromo-5-methoxyphenyl)ethanol ≥95%
1-(3-Bromo-5-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD28059764
CAS Number: 1823369-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR