2-Bromo-3-fluoro-4-methoxy-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

91920-500MG 91920-1G 91920-5G 91920-25G
77335-140EA 280.16 USD
77335-140 77359-232 77371-180 77391-660
2-Bromo-3-fluoro-4-methoxy-1-methylbenzene ≥95%
2-Bromo-3-fluoro-4-methoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD34772965
CAS Number: 2092621-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR