3-Bromo-2-fluoro-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

91919-500MG 91919-1G 91919-5G 91919-25G
77335-138EA 207.03 USD
77335-138 77359-230 77371-178 77391-658
3-Bromo-2-fluoro-4-methylphenol ≥97%
3-Bromo-2-fluoro-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD28739774
CAS Number: 1805554-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR