4-Bromo-N-methoxy-N,2-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

91883-500MG 91883-1G 91883-5G 91883-25G
77335-136EA 146.26 USD
77335-136 77345-186 77371-174 77391-656
4-Bromo-N-methoxy-N,2-dimethylbenzamide ≥97%
4-Bromo-N-methoxy-N,2-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11847399
CAS Number: 178313-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR