2,6-Dichlorothioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,6-Dichlorophenyl)(methyl)sulphane

91874-1G 91874-5G 91874-25G
77340-848EA 60.77 USD
77340-848 77371-170 77383-198
2,6-Dichlorothioanisole ≥97%
2,6-Dichlorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆Cl₂S
Storage Temperature: Ambient
MDL Number: MFCD03789170
CAS Number: 4210-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR