2,6-Dichloro-3-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

91872-1G 91872-5G 91872-25G
77340-846EA 60.77 USD
77340-846 77371-168 77383-196
2,6-Dichloro-3-(methylthio)benzaldehyde ≥97%
2,6-Dichloro-3-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂OS
MW: 221.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34772948
CAS Number: 2901109-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR