(2-Chloro-4,6-difluorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

91828-500MG 91828-1G 91828-5G
77336-322EA 386.25 USD
77336-322 77359-228 77383-192
(2-Chloro-4,6-difluorophenyl)(methyl)sulfane ≥95%
(2-Chloro-4,6-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅ClF₂S
MW: 194.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751304
CAS Number: 1805638-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR