2,3-Diethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

91761-500MG 91761-1G 91761-25G 91761-5G
77335-132EA 299.73 USD
77335-132 77359-226 77391-652 77383-190
2,3-Diethoxybenzaldehyde ≥95%
2,3-Diethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD05865164
CAS Number: 24454-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR