1-Bromo-2,3-diethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

91738-500MG 91738-1G 91738-5G
77340-014EA 712.76 USD
77340-014 77359-222 77383-186
1-Bromo-2,3-diethoxybenzene ≥95%
1-Bromo-2,3-diethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31591829
CAS Number: 2137677-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR