(2-Chloro-6-fluoro-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

91655-500MG 91655-1G 91655-5G
77339-954EA 692.16 USD
77339-954 77359-218 77383-182
(2-Chloro-6-fluoro-4-methylphenyl)methanol ≥97%
(2-Chloro-6-fluoro-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28751967
CAS Number: 1534389-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR