3-Methoxy-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

91601-500MG 91601-1G 91601-5G
77336-768EA 393.46 USD
77336-768 77359-216 77383-180
3-Methoxy-2,6-dimethylbenzaldehyde ≥97%
3-Methoxy-2,6-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16249778
CAS Number: 179554-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR