(3-Methoxy-2,6-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

91597-500MG 91597-1G 91597-5G
77335-130EA 347.11 USD
77335-130 77359-214 77383-178
(3-Methoxy-2,6-dimethylphenyl)boronic acid ≥97%
(3-Methoxy-2,6-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD18827124
CAS Number: 693285-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR