3,5-dimethyl-4-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

91200-500MG 91200-1G 91200-5G 91200-25G
77335-128EA 100.94 USD
77335-128 77342-968 77371-162 77383-174
3,5-dimethyl-4-(methylthio)phenol ≥97%
3,5-dimethyl-4-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₉H₁₂OS
Storage Temperature: Ambient
CAS Number: 7379-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR