1-Bromo-4-(methylsulfonyl)-2-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

91155-500MG 91155-1G 91155-5G 91155-25G
77335-126EA 196.73 USD
77335-126 77359-212 77371-160 77391-650
1-Bromo-4-(methylsulfonyl)-2-(trifluoromethyl)benzene ≥95%
1-Bromo-4-(methylsulfonyl)-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O₂S
MW: 303.096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527241
CAS Number: 1820711-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR