6-Bromo-2-fluoro-3-(methylsulfonyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

91149-500MG 91149-1G 91149-5G
77335-124EA 347.11 USD
77335-124 77359-210 77383-172
6-Bromo-2-fluoro-3-(methylsulfonyl)benzoic acid ≥97%
6-Bromo-2-fluoro-3-(methylsulfonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₄S
Storage Temperature: Ambient
CAS Number: 2918966-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR