(E)-3-bromo-2-chloro-6-fluorobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

91003-1G 91003-5G 91003-25G
77341-072EA 69.01 USD
77341-072 77371-154 77383-168
(E)-3-bromo-2-chloro-6-fluorobenzaldehyde oxime ≥97%
(E)-3-bromo-2-chloro-6-fluorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₄BrClFNO
Storage Temperature: Ambient
MDL Number: MFCD34772580
CAS Number: 2387488-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR