(E)-4-bromo-3,5-dimethylbenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

91001-1G 91001-5G 91001-25G
77341-516EA 90.64 USD
77341-516 77371-152 77383-164
(E)-4-bromo-3,5-dimethylbenzaldehyde oxime ≥97%
(E)-4-bromo-3,5-dimethylbenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD34772578
CAS Number: 2284536-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR