2-Chloro-6-fluoro-4-iodophenol ≥95%

Supplier: AOBChem USA

Warning

91000-500MG 91000-1G 91000-5G 91000-25G
77335-116EA 229.69 USD
77335-116 77359-206 77371-150 77391-646
2-Chloro-6-fluoro-4-iodophenol ≥95%
2-Chloro-6-fluoro-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃ClFIO
MW: 272.443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26895917
CAS Number: 1564859-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR