3-Fluoro-2-(methoxymethyl)-4-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

90767-500MG 90767-1G 90767-5G
77338-056EA 460.41 USD
77338-056 77359-202 77383-162
3-Fluoro-2-(methoxymethyl)-4-methylbenzaldehyde ≥95%
3-Fluoro-2-(methoxymethyl)-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34772458
CAS Number: 2918872-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR