1-Bromo-3-fluoro-2-(methoxymethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

90762-500MG 90762-1G 90762-5G 90762-25G
77335-112EA 264.71 USD
77335-112 77359-196 77371-146 77391-642
1-Bromo-3-fluoro-2-(methoxymethyl)-4-methylbenzene ≥97%
1-Bromo-3-fluoro-2-(methoxymethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34772453
CAS Number: 2901105-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR