1-(3-Fluorophenoxy)-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

90577-500MG 90577-1G 90577-5G 90577-25G
77335-108EA 199.82 USD
77335-108 77359-192 77371-142 77391-640
1-(3-Fluorophenoxy)-2-iodobenzene ≥97%
1-(3-Fluorophenoxy)-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₂H₈FIO
MW: 314.094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34772416
CAS Number: 1379658-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR