1-Bromo-2-(3-fluorophenoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

90575-1G 90575-5G 90575-25G
77341-646EA 97.85 USD
77341-646 77371-140 77383-150
1-Bromo-2-(3-fluorophenoxy)benzene ≥97%
1-Bromo-2-(3-fluorophenoxy)benzene

Precaución: for Research Use Only

Formula: C₁₂H₈BrFO
MW: 267.094 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25953914
CAS Number: 884512-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR