2-(3-Fluorophenoxy)aniline ≥97%
Supplier: AOBChem USA
|
Warning
|
90574-500MG
90574-1G
90574-5G
90574-25G
77335-106EA
150.38
USD
77335-106
77345-372
77371-138
77391-638
2-(3-Fluorophenoxy)aniline ≥97%
2-(3-Fluorophenoxy)aniline
Precaución: for Research Use Only
Formula:
C₁₂H₁₀FNO Storage Temperature: Ambient |
MDL Number:
MFCD04971079 CAS Number: 640766-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...