(4-(1,1-difluoroethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

90571-500MG 90571-1G 90571-25G 90571-5G
77335-102EA 300.76 USD
77335-102 77359-186 77391-634 77383-146
(4-(1,1-difluoroethyl)phenyl)(methyl)sulfane ≥95%
(4-(1,1-difluoroethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀F₂S
MW: 188.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34772415
CAS Number: 1895824-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR