(5-Methyl-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

90554-500MG 90554-1G 90554-5G 90554-25G
77335-100EA 121.54 USD
77335-100 77343-888 77371-136 77383-144
(5-Methyl-1,3-phenylene)bis(methylsulfane) ≥97%
(5-Methyl-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₁₂S₂
MW: 184.322 g/mol
Storage Temperature: Ambient
CAS Number: 2901066-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR