2-Bromo-4-iodo-5-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

90547-500MG 90547-1G 90547-5G
77336-988EA 402.73 USD
77336-988 77359-184 77383-142
2-Bromo-4-iodo-5-methoxypyridine ≥95%
2-Bromo-4-iodo-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrINO
MW: 313.918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34596139
CAS Number: 2415085-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR