3-(Cyclopropylmethoxy)-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

90034-500MG 90034-1G 90034-5G
77335-096EA 317.24 USD
77335-096 77359-182 77383-138
3-(Cyclopropylmethoxy)-5-methylbenzaldehyde ≥97%
3-(Cyclopropylmethoxy)-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD29122315
CAS Number: 1289090-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR