3-(Cyclopropylmethoxy)-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

90032-500MG 90032-1G 90032-5G 90032-25G
77335-094EA 242.05 USD
77335-094 77359-180 77371-132 77391-632
3-(Cyclopropylmethoxy)-5-methylbenzoic acid ≥97%
3-(Cyclopropylmethoxy)-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24500004
CAS Number: 1194258-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR