4-Bromo-2-(cyclopropylmethoxy)-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

90001-1G 90001-5G 90001-25G
77341-536EA 91.67 USD
77341-536 77371-130 77383-134
4-Bromo-2-(cyclopropylmethoxy)-1-(trifluoromethoxy)benzene ≥97%
4-Bromo-2-(cyclopropylmethoxy)-1-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34772095
CAS Number: 2918947-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR