4-((cyclopropylmethyl)thio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

89968-500MG 89968-1G 89968-5G 89968-25G
77335-088EA 271.92 USD
77335-088 77359-174 77371-128 77391-630
4-((cyclopropylmethyl)thio)benzoic acid ≥97%
4-((cyclopropylmethyl)thio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂S
MW: 208.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12138600
CAS Number: 1155956-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR