(4-Bromophenyl)(cyclopropylmethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

89966-500MG 89966-1G 89966-5G 89966-25G
77335-086EA 141.11 USD
77335-086 77344-954 77371-126 77391-628
(4-Bromophenyl)(cyclopropylmethyl)sulfane ≥97%
(4-Bromophenyl)(cyclopropylmethyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrS
Storage Temperature: Ambient
MDL Number: MFCD26396016
CAS Number: 917382-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR