4-Bromo-1-(cyclopropylmethoxy)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

89926-500MG 89926-1G 89926-5G 89926-25G
77335-080EA 160.68 USD
77335-080 77345-820 77371-118 77391-622
4-Bromo-1-(cyclopropylmethoxy)-2-methylbenzene ≥95%
4-Bromo-1-(cyclopropylmethoxy)-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD18294114
CAS Number: 1341978-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR