4-(Cyclopropylmethoxy)-3-fluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
89923-500MG
89923-1G
89923-5G
89923-25G
77335-078EA
114.33
USD
77335-078
77343-388
77371-116
77383-128
4-(Cyclopropylmethoxy)-3-fluorobenzoic acid ≥97%
4-(Cyclopropylmethoxy)-3-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₁FO₃ MW: 210.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11156579 CAS Number: 1021144-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...