4-Bromo-1-(cyclopropylmethoxy)-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

89920-500MG 89920-1G 89920-5G 89920-25G
77335-076EA 99.91 USD
77335-076 77342-906 77371-114 77383-126
4-Bromo-1-(cyclopropylmethoxy)-2-fluorobenzene ≥97%
4-Bromo-1-(cyclopropylmethoxy)-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD28676026
CAS Number: 622386-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR