3,5-Dibromo-4-chlorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

89881-500MG 89881-1G 89881-5G
77336-918EA 399.64 USD
77336-918 77359-168 77383-118
3,5-Dibromo-4-chlorobenzaldehyde ≥95%
3,5-Dibromo-4-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Br₂ClO
MW: 298.359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32061830
CAS Number: 861303-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR