Methyl 3,5-dibromo-4-chlorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
89878-500MG
89878-1G
89878-5G
89878-25G
77335-070EA
263.68
USD
77335-070
77359-166
77371-106
77391-620
Methyl 3,5-dibromo-4-chlorobenzoate ≥97%
Methyl 3,5-dibromo-4-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₅Br₂ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD29918460 CAS Number: 1839608-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...