1-Bromo-2-fluoro-3-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

89523-500MG 89523-1G 89523-5G 89523-25G
77335-066EA 187.46 USD
77335-066 77359-162 77371-102 77391-618
1-Bromo-2-fluoro-3-methoxy-4-methylbenzene ≥97%
1-Bromo-2-fluoro-3-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771881
CAS Number: 2092391-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR