(4-(Benzyloxy)naphthalen-1-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

89522-500MG 89522-1G 89522-5G 89522-25G
77335-064EA 244.11 USD
77335-064 77359-160 77371-100 77391-616
(4-(Benzyloxy)naphthalen-1-yl)(methyl)sulfane ≥97%
(4-(Benzyloxy)naphthalen-1-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₈H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD34771880
CAS Number: 2901102-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR