1-(2,3-Difluorophenyl)-4,4-Dimethylcyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89430-500MG 89430-1G 89430-5G 89430-25G
77335-060EA 202.91 USD
77335-060 77359-156 77371-096 77391-612
1-(2,3-Difluorophenyl)-4,4-Dimethylcyclohexanol ≥95%
1-(2,3-Difluorophenyl)-4,4-Dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₄H₁₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD21392344
CAS Number: 1484538-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR