1-(2-Ethoxyphenyl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89417-500MG 89417-1G 89417-5G 89417-25G
77335-058EA 243.08 USD
77335-058 77359-154 77371-094 77391-610
1-(2-Ethoxyphenyl)-4,4-dimethylcyclohexanol ≥97%
1-(2-Ethoxyphenyl)-4,4-dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₆H₂₄O₂
MW: 248.361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20166520
CAS Number: 1491495-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR