1-(5-Bromo-2-fluorophenyl)-4,4-dimethylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89411-500MG 89411-1G 89411-5G 89411-25G
77335-056EA 243.08 USD
77335-056 77359-152 77371-092 77391-608
1-(5-Bromo-2-fluorophenyl)-4,4-dimethylcyclohexanol ≥97%
1-(5-Bromo-2-fluorophenyl)-4,4-dimethylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrFO
MW: 301.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23340415
CAS Number: 1503788-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR