1-(5-Bromo-2-fluorophenyl)-4-methylcyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89310-500MG 89310-1G 89310-5G 89310-25G
77335-054EA 182.31 USD
77335-054 77347-062 77371-090 77391-606
1-(5-Bromo-2-fluorophenyl)-4-methylcyclohexanol ≥97%
1-(5-Bromo-2-fluorophenyl)-4-methylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD23759084
CAS Number: 1505414-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR