1-(4-Fluoro-3-(trifluoromethyl)phenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89236-1G 89236-5G 89236-25G
77340-844EA 60.77 USD
77340-844 77371-086 77383-108
1-(4-Fluoro-3-(trifluoromethyl)phenyl)cyclohexanol ≥97%
1-(4-Fluoro-3-(trifluoromethyl)phenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₄F₄O
MW: 262.243 g/mol
Storage Temperature: Ambient
CAS Number: 1933438-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR